accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421261-82-6

NNNS

CAS RN 1421261-82-6

IUPAC Name: 1H,3H-thieno[3,4-b]quinoxaline-6-carbonitrile
Synonyms: 1H,3H-thieno[3,4-b]quinoxaline-6-carbonitrile
Molecular Weight (g/mol): 213.26
Molecular Formula: C11H7N3S
InChi Key: RHAQHNRQPSWJRH-UHFFFAOYSA-N
SMILES: N#CC1=CC2=NC3=C(CSC3)N=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results