accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHSOONHIF

CAS RN 1421261-99-5

IUPAC Name: N-{4-[(2-fluoro-4-iodophenyl)sulfamoyl]phenyl}acetamide
Synonyms: N-{4-[(2-fluoro-4-iodophenyl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 434.22
Molecular Formula: C14H12FIN2O3S
InChi Key: MSFKCXKKEQSELC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=C(F)C=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results