accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHClClBr

CAS RN 1421262-30-7

IUPAC Name: 2-bromo-N-[(3,4-dichlorophenyl)methyl]benzene-1-sulfonamide
Synonyms: 2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide
Molecular Weight (g/mol): 395.09
Molecular Formula: C13H10BrCl2NO2S
InChi Key: BBKPFBULBLAWNE-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C(CNS(=O)(=O)C2=CC=CC=C2Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results