accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNHONN

CAS RN 1421263-07-1

IUPAC Name: 6-oxo-5-(thiophen-3-yl)-1,6-dihydropyrazine-2,3-dicarbonitrile
Synonyms: 5-oxo-6-(thiophen-3-yl)-4H-pyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 228.23
Molecular Formula: C10H4N4OS
InChi Key: UNKZQYRUAFQDJO-UHFFFAOYSA-N
SMILES: O=C1NC(C#N)=C(N=C1C1=CSC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results