accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNNOOBr

CAS RN 1421263-13-9

IUPAC Name: 3-(4-bromophenyl)-5-methyl-4-(3-nitrophenyl)-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-5-methyl-4-(3-nitrophenyl)-1,2,4-triazole
Molecular Weight (g/mol): 359.18
Molecular Formula: C15H11BrN4O2
InChi Key: WQZPCZDVKCPFRR-UHFFFAOYSA-N
SMILES: CC1=NN=C(N1C1=CC(=CC=C1)[N+]([O-])=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)