accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421263-46-8

NNSN

CAS RN 1421263-46-8

IUPAC Name: 4-{2-[(pyridin-2-yl)methyl]-1,3-thiazol-4-yl}pyridine
Synonyms: 4-[2-(pyridin-2-ylmethyl)-1,3-thiazol-4-yl]pyridine
Molecular Weight (g/mol): 253.32
Molecular Formula: C14H11N3S
InChi Key: RFXGELSJVHDCRI-UHFFFAOYSA-N
SMILES: C(C1=NC(=CS1)C1=CC=NC=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results