accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421263-82-2

IUPAC Name: 3-{[(6-cyanopyridin-3-yl)[(2-cyanopyridin-3-yl)methyl]amino]methyl}pyridine-2-carbonitrile
Synonyms: 3-{[(6-cyanopyridin-3-yl)[(2-cyanopyridin-3-yl)methyl]amino]methyl}pyridine-2-carbonitrile
Molecular Weight (g/mol): 351.37
Molecular Formula: C20H13N7
InChi Key: MGYJYXRHCRJQQV-UHFFFAOYSA-N
SMILES: N#CC1=NC=C(C=C1)N(CC1=C(N=CC=C1)C#N)CC1=C(N=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results