CAS RN 1421773-36-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Boc-aminomethyl)-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1421773-36-5 Synonym: 3-(Boc-aminomethyl)-5-fluorophenylboronic acid pinacol ester