accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14220-21-4

IUPAC Name: bromorhenium; pentakis(methanidylidyneoxidanium)
Synonyms: bromopentacarbonylrhenium(I)
Molecular Weight (g/mol): 406.16
Molecular Formula: C5BrO5Re
InChi Key: NWJBOTGGBYFKEJ-UHFFFAOYSA-M
SMILES: Br[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results