Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Methyl (S)-3-Bromo-2-Methylpropionate

Catalog No. 3249225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-492-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 32-492-25G Supplier Sigma Aldrich Supplier No. 3249225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Methyl (S)-3-bromo-2-methylpropionate can be used:As a reference substrate in the alkylation reactions of vitamin B12 derivatives.As a starting material in the preparation of biologically significant 3-[(2H-1,2,4-benzothiadiazine-1,1-dioxide-3-yl)thio]-2-methylpropanoic acids.

Specifications

CAS 14220-21-4
Molecular Formula C5BrO5Re
Refractive Index n20/D 1.455
Linear Formula Re(CO)5Br
Quantity 5 g
MDL Number MFCD00013295
Synonym (-)-Methyl (S)-beta-bromoisobutyrate
Molecular Weight (g/mol) 406.16 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.