CAS RN 14221-01-3
Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis)
CAS: 14221-01-3 Molecular Formula: C72H60P4Pd Molecular Weight (g/mol): 1155.59 MDL Number: MFCD00010012 InChI Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N Synonym: tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4 PubChem CID: 11979704 IUPAC Name: palladium;triphenylphosphane SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Tetrakis(triphenylphosphine)palladium(0), 99%
CAS: 14221-01-3 Molecular Formula: C72H60P4Pd Molecular Weight (g/mol): 1155.59 MDL Number: MFCD00010012 InChI Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N Synonym: tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4 PubChem CID: 11979704 IUPAC Name: palladium;triphenylphosphane SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min
CAS: 14221-01-3 Molecular Formula: C72H60P4Pd Molecular Weight (g/mol): 1155.59 MDL Number: MFCD00010012 InChI Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N PubChem CID: 11979704 SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Tetrakis(triphenylphosphine)palladium(0) 97.0+%, TCI America™
CAS: 14221-01-3 Molecular Formula: C72H60P4Pd Molecular Weight (g/mol): 1155.59 MDL Number: MFCD00010012 InChI Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N Synonym: tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4 PubChem CID: 11979704 IUPAC Name: tetrakis(triphenylphosphane) palladium SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Tetrakis(triphenylphosphine)palladium(0), Approx. 99%, Spectrum™ Chemical
CAS: 14221-01-3 Molecular Formula: C72H60P4Pd Molecular Weight (g/mol): 1155.59 MDL Number: MFCD00010012 InChI Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N IUPAC Name: tetrakis(triphenylphosphane) palladium SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Sigma Aldrich Tetrakis(Triphenylphosphine)Palladium(0)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetrakis(Triphenylphosphine)Palladium(0)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.