Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(triphenylphosphine)palladium(0), Approx. 99%, Spectrum™ Chemical
SDP

Catalog No. 18607336 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
200 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-336 25 g Amber Glass Bottle
18-607-333 1 g Amber Glass Bottle
18-607-334 5 g Amber Glass Bottle
18-607-335 200 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-336 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T202025GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T2020, 14221-01-3, C72H60P4Pd

Spectrum™ Chemical Tetrakis(triphenylphosphine)palladium(0)

Chemical Identifiers

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Molecular Weight (g/mol) 1155.59
MDL Number MFCD00010012
InChI Key NFHFRUOZVGFOOS-UHFFFAOYSA-N
IUPAC Name tetrakis(triphenylphosphane) palladium
SMILES [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity ∼99%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.