accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142214-04-8

IUPAC Name: rhodium(3+) tritriphenylacetate
Synonyms: rhodium(3+) tritriphenylacetate
Molecular Weight (g/mol): 964.92
Molecular Formula: C60H45O6Rh
InChi Key: LJIVYHNRWSWCRU-UHFFFAOYSA-K
SMILES: [Rh+3].[O-]C(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results