Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-2-Ethoxyvinylboronic Acid Pinacol Ester

Catalog No. 7315281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-152-81G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73-152-81G Supplier Sigma Aldrich Supplier No. 7315281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-2-Ethoxyvinylboronic acid pinacol ester is a boronic ester commonly used in Suzuki-Miyaura cross-coupling.This reaction is a key step to synthesize:Azaindole and diazaindoles from chloroamino-N-heterocycles.Doryanine and its derivatives from 2-bromobenzoic acid.

Specifications

CAS 142214-04-8
Melting Point 260°C to 263°C
Molecular Formula C80H60O8Rh2 -+ CH2Cl2
Refractive Index n20/D 1.447
Linear Formula C80H60O8Rh2 -+ CH2Cl2
MDL Number MFCD15071082
Quantity 100 mg
Synonym (E)-2-(2-Ethoxyvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; (E)-2-Ethoxyvinylboronic acid pinacol ester
Molecular Weight (g/mol) 1440.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.