accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONN

CAS RN 142253-54-1

IUPAC Name: tert-butyl 3-cyanoazetidine-1-carboxylate
Synonyms: tert-butyl 3-cyanoazetidine-1-carboxylate
Molecular Weight (g/mol): 182.22
Molecular Formula: C9H14N2O2
InChi Key: WEFREESWPHICPL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results