Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-3-cyanoazetidine, 97%, Thermo Scientific™

Catalog No. AAH2884103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2884103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2884103 Supplier Thermo Scientific Chemicals Supplier No. H2884103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142253-54-1
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.22
MDL Number MFCD06796640
InChI Key WEFREESWPHICPL-UHFFFAOYSA-N
Synonym 1-boc-3-cyanoazetidine, 1-n-boc-3-cyano-azetidine, 1-n-boc-3-cyanoazetidine, 3-cyano-azetidine-1-carboxylic acid tert-butyl ester, 3-cyano-1-boc-azetidine, n-boc-3-cyanoazetidine, n-t-boc-3-cyanoazetidine, 1-n-boc-3-cyano azetidine, tert-butyl 3-cyanoazetidinecarboxylate
PubChem CID 10631283
IUPAC Name tert-butyl 3-cyanoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C#N

Specifications

Melting Point 70°C to 71°C
UN Number UN3439
Quantity 1 g
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 1-Boc-3-cyanoazetidine
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.