CAS RN 1423715-09-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ LSD1 Inhibitor VII, SP-2509
A cell-permeable, potent, selective, reversible inhibitor of Lysine-Specific Demethylase 1 (LSD1; IC50 = 13 nM; Ki = 31 nM).