accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNOOCH3

CAS RN 142593-07-5

IUPAC Name: 6,6'-dimethyl [2,2'-bipyridine]-6,6'-dicarboxylate
Synonyms: 6,6'-dimethyl [2,2'-bipyridine]-6,6'-dicarboxylate
Molecular Weight (g/mol): 272.26
Molecular Formula: C14H12N2O4
InChi Key: YFKCGCGCKIYMIH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=N1)C1=NC(=CC=C1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results