Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate 95.0+%, TCI America™
SDP

Catalog No. D5025200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5025200MG 200 mg
D50251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5025200MG Supplier TCI America Supplier No. D5025200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142593-07-5
Molecular Formula C14H12N2O4
Molecular Weight (g/mol) 272.26
MDL Number MFCD11042478
InChI Key YFKCGCGCKIYMIH-UHFFFAOYSA-N
Synonym 2,2′C-Bipyridine-6,6′C-dicarboxylic Acid Dimethyl Ester, 6,6′C-Bis(methoxycarbonyl)-2,2′C-bipyridine, 6,6′C-Dimethoxycarbonyl-2,2′C-bipyridine
PubChem CID 11477388
IUPAC Name methyl 6-(6-methoxycarbonylpyridin-2-yl)pyridine-2-carboxylate
SMILES COC(=O)C1=CC=CC(=N1)C2=NC(=CC=C2)C(=O)OC

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 200 mg
Formula Weight 272.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,2′-Bipyridine-6,6′-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.