accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOHOONNH(S)(S)H3CH3C(R)CH3H3CH3CCH3

CAS RN 1425938-63-1

IUPAC Name: (4S)-3-[(2S,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S)-3-[(2S,3R)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 524.61
Molecular Formula: C29H36N2O7
InChi Key: ZWRFHHZZRMZKFK-CQLNOVPUSA-N
SMILES: C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)N1[C@@H](COC1(C)C)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)