accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142600-61-1

IUPAC Name: λ²-cobalt(2+) bis(2-methoxyethan-1-olate)
Synonyms: λ²-cobalt(2+) bis(2-methoxyethanolate)
Molecular Weight (g/mol): 209.11
Molecular Formula: C6H14CoO4
InChi Key: BFBNUELICQBJBC-UHFFFAOYSA-N
SMILES: [Co++].COCC[O-].COCC[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results