CAS RN 14263-94-6
CAS RN 14263-94-6
IUPAC Name:
zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
Synonyms:
zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
Molecular Weight (g/mol):
475.46
Molecular Formula:
C14H12Cl4N4O2Zn
InChi Key:
GPPKNJIWDULNQH-UHFFFAOYSA-J
SMILES:
[Cl-].[Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(=CC=C1[N+]#N)C1=CC=C([N+]#N)C(OC)=C1
1
–
1
of
1
results
1
MP Biomedicals, Inc Fast Blue B Salt, MP Biomedicals™
CAS: 14263-94-6 Molecular Formula: C14H16Cl4N4O2Zn+2 Molecular Weight (g/mol): 479.488 InChI Key: ZZJFLKSMXLBSAQ-UHFFFAOYSA-N Synonym: Azoic Diazo No. 48, Diazo Blue B, o-Dianisidine bis(diazotized) zinc double salt, Naphthanil Diazo Blue B, DBB PubChem CID: 129894309 IUPAC Name: 4-(4-diazonio-3-methoxyphenyl)-2-methoxybenzenediazonium;zinc;tetrahydrochloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]#N)OC)[N+]#N.Cl.Cl.Cl.Cl.[Zn]