Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Blue B Salt, MP Biomedicals™

Catalog No. MP215020125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215020125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215020125 Supplier MP Biomedicals, Inc Supplier No. 0215020125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a tissue stain. Fast Blue B Salt, MP Biomedicals is useful for TLC of Aflatoxins.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14263-94-6
Molecular Formula C14H16Cl4N4O2Zn+2
Molecular Weight (g/mol) 479.488
InChI Key ZZJFLKSMXLBSAQ-UHFFFAOYSA-N
Synonym Azoic Diazo No. 48, Diazo Blue B, o-Dianisidine bis(diazotized) zinc double salt, Naphthanil Diazo Blue B, DBB
PubChem CID 129894309
IUPAC Name 4-(4-diazonio-3-methoxyphenyl)-2-methoxybenzenediazonium;zinc;tetrahydrochloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]#N)OC)[N+]#N.Cl.Cl.Cl.Cl.[Zn]

Specifications

Solubility Information Soluble in water (4mg/4mL, clear solution).
Quantity 25 g
Physical Form Powder
Formula Weight 475.47
Recommended Storage 2–8°C. Protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.