accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHN(S)H2N

CAS RN 142643-29-6

IUPAC Name: (3S)-3-({[(tert-butoxy)carbonyl]amino}methyl)piperidin-1-ium
Synonyms: (3S)-3-{[(tert-butoxycarbonyl)amino]methyl}piperidin-1-ium
Molecular Weight (g/mol): 215.32
Molecular Formula: C11H23N2O2
InChi Key: KHPQHXGYYXYTDN-SECBINFHSA-O
SMILES: CC(C)(C)OC(=O)NC[C@@H]1CCC[NH2+]C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)