accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCl

CAS RN 14273-86-0

IUPAC Name: methyl 5-chloropentanoate
Synonyms: methyl 5-chloropentanoate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: JAVHFVJOWIQHII-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)