Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-chloropentanoate, 97%, Thermo Scientific™

Catalog No. AC278251000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC278251000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC278251000 Supplier Thermo Scientific Chemicals Supplier No. 278251000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14273-86-0
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00065274
InChI Key JAVHFVJOWIQHII-UHFFFAOYSA-N
Synonym methyl 5-chlorovalerate, methyl .delta.-chlorovalerate, clch2 ch2 3c o och3, acmc-20akg1, methyl delta-chlorovalerate, methyl 5-chloranylpentanoate, 5-chlorovaleric acid methyl ester, 5-chlorovaleric acid, methyl ester, 5-chloropentanoic acid methyl ester
PubChem CID 274413
IUPAC Name methyl 5-chloropentanoate
SMILES COC(=O)CCCCCl

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4378 to 1.4398 (20°C, 589nm)
Linear Formula Cl(CH2)4C(O)OCH3
Quantity 100 g
Formula Weight 150.6
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Methyl 5-chloropentanoate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.