accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 14273-90-6

IUPAC Name: methyl 6-bromohexanoate
Synonyms: methyl 6-bromohexanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: KYLVAMSNNZMHSX-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)