Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-bromohexanoate, 96+%

Catalog No. AC392691000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392691000 100 g Glass bottle
AC392690050 5 g Glass bottle
AC392690250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392691000 Supplier Thermo Scientific Chemicals Supplier No. 392691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14273-90-6 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14273-90-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00187528
InChI Key KYLVAMSNNZMHSX-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid methyl ester, hexanoic acid, 6-bromo-, methyl ester, methyl-6-bromohexanoate, methyl6-bromohexanoate, pubchem16910, methyl 6-bromo-hexanoate, 6-bromo-hexanoic acid methyl ester, 6-bromo-hexanoic acid-methyl ester, 6-bromo-henxanoic acid methyl ester, hexanoic acid,6-bromo-, methyl ester
PubChem CID 4170292
IUPAC Name methyl 6-bromohexanoate
SMILES COC(=O)CCCCCBr

Specifications

Color Colorless to Yellow
Density 1.3160g/mL
Boiling Point 150.0°C (50.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4630 to 1.4650
Linear Formula Br(CH2)5C(O)OCH3
Quantity 100 g
Specific Gravity 1.316
Formula Weight 209.08
Percent Purity 96+%
Physical Form Liquid
Chemical Name or Material Methyl 6-bromohexanoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.