accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142753-10-4

IUPAC Name: [2-(1,3-dioxolan-2-yl)ethyl](methyl)amine
Synonyms: [2-(1,3-dioxolan-2-yl)ethyl](methyl)amine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ZUUQKDDUUOFCNP-UHFFFAOYSA-N
SMILES: CNCCC1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results