accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14282-91-8

IUPAC Name: ruthenium(3+) hexaamine trichloride
Synonyms: ruthenium(3+) hexaamine trichloride
Molecular Weight (g/mol): 309.61
Molecular Formula: Cl3H18N6Ru
InChi Key: GBDZMMXUOBAJMN-UHFFFAOYSA-K
SMILES: N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (6)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)