Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexaammineruthenium(III) Chloride

Catalog No. 503081468 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1468 1 g
26-200-550G 50 g
26-200-55G 5 g
26-200-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 50-308-1468 Supplier Sigma Aldrich Supplier No. 2620051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexaammineruthenium III/II (HexRu(III)/HexRu(II)) couple is a commonly used electrochemical redox couple due to its chemical and electrochemical reversibility.Hexaammineruthenium(III) chloride can be used to synthesize chloropentaammineruthenium(III) chloride.

Specifications

CAS 14282-91-8
Molecular Formula H18Cl3N6Ru
Linear Formula [Ru(NH3)6]Cl3
Quantity 1 g
MDL Number MFCD00011478
Molecular Weight (g/mol) 309.61 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.