CAS RN 1429340-48-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
CAS: 1429340-48-6 Synonym: 2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester