accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143-08-8

IUPAC Name: nonan-1-ol
Synonyms: nonan-1-ol alcohol C-9 nonylalkohol C9 alcohol fatty alcohol(C9) pelargonic alcohol pelargonalkohol nonalol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: ZWRUINPWMLAQRD-UHFFFAOYSA-N
SMILES: CCCCCCCCCO
Molecular Weight (g/mol) 
  • (4)
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (1)
  • (2)
  • (5)
  • (4)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (1)