Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonanol, MP Biomedicals

Catalog No. ICN216403 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216403 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216403 Supplier MP Biomedicals, Inc Supplier No. 0521640380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143-08-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
InChI Key ZWRUINPWMLAQRD-UHFFFAOYSA-N
Synonym 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
PubChem CID 8914
ChEBI CHEBI:35986
IUPAC Name nonan-1-ol
SMILES CCCCCCCCCO

Specifications

Quantity 100 g
Formula Weight 144.26
Chemical Name or Material 1-Nonanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.