CAS RN 143-18-0
Potassium Oleate 98.0+%, TCI America™
CAS: 143-18-0 Molecular Formula: C18H33KO2 Molecular Weight (g/mol): 320.558 MDL Number: MFCD00064243 InChI Key: MLICVSDCCDDWMD-UHFFFAOYSA-M Synonym: acmc-209cpw,9-octadecenoic acid 9z-, potassium salt 1:1,ksc175g6j PubChem CID: 44135690 IUPAC Name: potassium;octadec-9-enoate SMILES: CCCCCCCCC=CCCCCCCCC(=O)[O-].[K+]
MP Biomedicals, Inc Potassium Oleate, MP Biomedicals
CAS: 143-18-0 Molecular Formula: C18H33KO2 Molecular Weight (g/mol): 320.558 InChI Key: MLICVSDCCDDWMD-UHFFFAOYSA-M Synonym: acmc-209cpw,9-octadecenoic acid 9z-, potassium salt 1:1,ksc175g6j PubChem CID: 44135690 IUPAC Name: potassium;octadec-9-enoate SMILES: CCCCCCCCC=CCCCCCCCC(=O)[O-].[K+]