Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oleate, MP Biomedicals

Catalog No. ICN217101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217101 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217101 Supplier MP Biomedicals, Inc Supplier No. 0521710125

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143-18-0
Molecular Formula C18H33KO2
Molecular Weight (g/mol) 320.558
InChI Key MLICVSDCCDDWMD-UHFFFAOYSA-M
Synonym acmc-209cpw, 9-octadecenoic acid 9z-, potassium salt 1:1, ksc175g6j
PubChem CID 44135690
IUPAC Name potassium;octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)[O-].[K+]

Specifications

Quantity 100 g
Formula Weight 320.56
Chemical Name or Material Potassium Oleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.