CAS RN 143-28-2
Oleyl alcohol, 99+%
CAS: 143-28-2 Molecular Formula: C18H36O Molecular Weight (g/mol): 268.49 MDL Number: MFCD00002993 InChI Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N Synonym: oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol PubChem CID: 5284499 ChEBI: CHEBI:73504 IUPAC Name: (Z)-octadec-9-en-1-ol SMILES: CCCCCCCC\C=C/CCCCCCCCO
Oleyl Alcohol, Approx. 99%, MP Biomedicals™
CAS: 143-28-2 Molecular Formula: C18H36O Molecular Weight (g/mol): 268.49 MDL Number: MFCD00002993 InChI Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N Synonym: oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol PubChem CID: 5284499 ChEBI: CHEBI:73504 IUPAC Name: (9Z)-octadec-9-en-1-ol SMILES: CCCCCCCC\C=C/CCCCCCCCO
Oleyl alcohol, ca. 60%, technical
CAS: 143-28-2 Molecular Formula: C18H36O Molecular Weight (g/mol): 268.49 MDL Number: MFCD00002993 InChI Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N Synonym: oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol PubChem CID: 5284499 ChEBI: CHEBI:73504 IUPAC Name: (9Z)-octadec-9-en-1-ol SMILES: CCCCCCCC\C=C/CCCCCCCCO
Oleyl alcohol, tech. 80-85%, Thermo Scientific Chemicals
CAS: 143-28-2 Molecular Formula: C18H36O Molecular Weight (g/mol): 268.49 MDL Number: MFCD00002993 InChI Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N Synonym: oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol PubChem CID: 5284499 ChEBI: CHEBI:73504 IUPAC Name: (9Z)-octadec-9-en-1-ol SMILES: CCCCCCCC\C=C/CCCCCCCCO
Oleyl Alcohol 60.0+%, TCI America™
CAS: 143-28-2 Molecular Formula: C18H36O Molecular Weight (g/mol): 268.49 MDL Number: MFCD00002993 InChI Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N Synonym: oleyl alcohol,cis-9-octadecen-1-ol,z-octadec-9-en-1-ol,ocenol,conditioner 1,dermaffine,lancol,novol,oceol,oleol PubChem CID: 5284499 ChEBI: CHEBI:73504 IUPAC Name: (9Z)-octadec-9-en-1-ol SMILES: CCCCCCCC\C=C/CCCCCCCCO
Sigma Aldrich Oleyl alcohol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.