Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl Alcohol, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013111 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3111 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3111 Supplier MilliporeSigma Supelco Supplier No. 08066100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 143-28-2
Density 0.849 g/mL (at 25°C)
Linear Formula CH3(CH2)7 CH=CH(CH2)7 CH2OH
MDL Number MFCD00002993
Melting Point 0°C to 5.0°C
Boiling Point 207°C (13 mmHg)
Synonym Z-9-Dodecen-1-ol; cis-9-Octadecen-1-ol
Quantity 100 mg
Formula Weight 268.48
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.