accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143-36-2

IUPAC Name: iodo(methyl)mercury
Synonyms: iodo(methyl)mercury
Molecular Weight (g/mol): 342.53
Molecular Formula: CH3HgI
InChi Key: JVDIOYBHEYUIBM-UHFFFAOYSA-M
SMILES: C[Hg]I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results