Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmercury(II) iodide, 98+%, Thermo Scientific™

Catalog No. AA3712804
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3712804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3712804 Supplier Thermo Scientific Chemicals Supplier No. 3712804

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-36-2
Molecular Formula CH3HgI
Molecular Weight (g/mol) 342.53
MDL Number MFCD00013701
InChI Key JVDIOYBHEYUIBM-UHFFFAOYSA-M
Synonym methylmercuric iodide, mercury, iodomethyl, methyliodomercury, methylmercury ii iodide, iodo methyl mercury, methylmercury iodide, methyl mercuric iodide, methyl mercury ii iodide, mercury, iodomethyl-8ci 9ci, mercury, iodomethyl-7ci,8ci,9ci
PubChem CID 67347
IUPAC Name iodo(methyl)mercury
SMILES C[Hg]I

Specifications

Melting Point 143°C
Quantity 2 g
UN Number UN2025
Formula Weight 342.56
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Methylmercury(II) iodide
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.