accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClOClClClClCl

CAS RN 143-50-0

IUPAC Name: decachloropentacyclo[5.3.0.0²,⁶.0³,⁹.0⁴,⁸]decan-5-one
Synonyms: kepone chlordecone
Molecular Weight (g/mol): 490.61
Molecular Formula: C10Cl10O
InChi Key: LHHGDZSESBACKH-UHFFFAOYNA-N
SMILES: ClC12C(=O)C3(Cl)C4(Cl)C1(Cl)C1(Cl)C2(Cl)C3(Cl)C4(Cl)C1(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results