CAS RN 143-74-8
Thermo Scientific Chemicals Phenol Red, ACS
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonphthalein; Phenylsulfonephthalein PubChem CID: 4766 ChEBI: CHEBI:31991 SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenol Red, pure, indicator
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonephthalein PubChem CID: 4766 ChEBI: CHEBI:31991 SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenol Red
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: phenol red,phenolsulfonphthalein,fenolipuna,sulfonphthal,sulphental,sulphonthal,phenolsulphonphthalein,psp indicator,phenolsulfonephthalein,unii-i6g9y0j1oj PubChem CID: 4766 ChEBI: CHEBI:31991 SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenol Red, ACS reagent
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonephthalein PubChem CID: 4766 ChEBI: CHEBI:31991 SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol red free acid, ACS reagent grade, MP Biomedicals™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonephthalein sodium salt PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red (0.1% in ca. 20% Ethanol) [for Titration], TCI America™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: phenol red,phenolsulfonphthalein,fenolipuna,sulfonphthal,sulphental,sulphonthal,phenolsulphonphthalein,psp indicator,phenolsulfonephthalein,unii-i6g9y0j1oj PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red, TCI America™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonphthalein PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red (0.04% in Water) [for pH Determination], TCI America™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: phenol red,phenolsulfonphthalein,fenolipuna,sulfonphthal,sulphental,sulphonthal,phenolsulphonphthalein,psp indicator,phenolsulfonephthalein,unii-i6g9y0j1oj PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red, ACS Reagent (Phenolsulfonphthalein), Reagents
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonphthalein IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenol Red Indicator, ACS Reagent, Solstice
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.38 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonphthalein PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
- 1
- 2