accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143085-87-4

IUPAC Name: 3-methyl-N-(4-phenylbutan-2-yl)butanamide
Synonyms: 3-methyl-N-(4-phenylbutan-2-yl)butanamide
Molecular Weight (g/mol): 233.36
Molecular Formula: C15H23NO
InChi Key: KIJQPTRIOXCOEG-UHFFFAOYNA-N
SMILES: CC(C)CC(=O)NC(C)CCC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results