Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Methyl-3-phenylpropyl)isovaleramide, TCI America™
SDP

Catalog No. M097310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M097310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M097310G Supplier TCI America Supplier No. M097310G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143085-87-4
Molecular Formula C15H23NO
Molecular Weight (g/mol) 233.355
MDL Number MFCD00144820
InChI Key KIJQPTRIOXCOEG-UHFFFAOYSA-N
Synonym 3-Isovaleramido-1-phenylbutane
PubChem CID 4070752
IUPAC Name 3-methyl-N-(4-phenylbutan-2-yl)butanamide
SMILES CC(C)CC(=O)NC(C)CCC1=CC=CC=C1

Specifications

Melting Point 52°C
Color Yellow
Quantity 10 g
Formula Weight 233.36
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Methyl-3-phenylpropyl)isovaleramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.