accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3

CAS RN 14315-97-0

IUPAC Name: 1,1,3-trimethoxypropane
Synonyms: 1,1,3-trimethoxypropane
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: FKZYYYDRLJCHGL-UHFFFAOYSA-N
SMILES: COCCC(OC)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)