accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(R)H3CNH2HCl

CAS RN 14316-06-4

IUPAC Name: hydrogen methyl 2-aminopropanoate chloride
Synonyms: hydrogen methyl 2-aminopropanoate chloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(C)N
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (7)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (5)
  • (4)