Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Alanine methyl ester hydrochloride, 98%

Catalog No. AC435320250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435320250 25 g Glass bottle
AC435320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435320250 Supplier Thermo Scientific Chemicals Supplier No. 435320250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14316-06-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14316-06-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00066141
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
PubChem CID 11182647
IUPAC Name methyl (2R)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(C)N

Specifications

Melting Point 60.4°C to 62.1°C
Color White
Elemental Analysis 33.56 to 35.28% C, 9.78 to 10.28% N
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition - 12.50 (19.00°C +/-2.5,c=2,MeOH)
Specific Rotation -12.5
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine methyl ester hydrochloride
Show More Show Less
EINECSNumber 238-258-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.