accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOO

CAS RN 14316-61-1

IUPAC Name: 2-phenoxyacetyl 2-phenoxyacetate
Synonyms: 2-phenoxyacetyl 2-phenoxyacetate
Molecular Weight (g/mol): 286.28
Molecular Formula: C16H14O5
InChi Key: CCSBNBKMACZDGN-UHFFFAOYSA-N
SMILES: O=C(COC1=CC=CC=C1)OC(=O)COC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results