Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxyacetic Anhydride 98.0+%, TCI America™
SDP

Catalog No. P011125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011125G 25 g
P0111250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P011125G Supplier TCI America Supplier No. P011125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14316-61-1
Molecular Formula C16H14O5
Molecular Weight (g/mol) 286.283
MDL Number MFCD00059681
InChI Key CCSBNBKMACZDGN-UHFFFAOYSA-N
PubChem CID 318938
IUPAC Name (2-phenoxyacetyl) 2-phenoxyacetate
SMILES C1=CC=C(C=C1)OCC(=O)OC(=O)COC2=CC=CC=C2

Specifications

Melting Point 73°C
Color White
Quantity 25 g
Formula Weight 286.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxyacetic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.