accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143218-48-8

IUPAC Name: sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate
Synonyms: sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate
Molecular Weight (g/mol): 214.59
Molecular Formula: C3H8ClNaO5S
InChi Key: ZPFGAXXLEFTBEU-UHFFFAOYNA-M
SMILES: O.[Na+].OC(CCl)CS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results